Sodium tetradecyl sulphate 95% cont. up to ca 5% sodium methyl sulfate
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Tetradecyl sulphuric acid salt, Tetradecyl sulphate sodium, Sodium 1-tetradecyl sulphate
CAS Number:139-88-8
Formula:C₁₄H₂₉NaO₄S
MDL Number:MFCD00007468
:316.44 g/mol
Merck Index:12,08838
Melting Pt:182…192 °C
Storage Temperature:Ambient
EINECS:214-737-2
UN:3267
ADR:8,III
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together