Order Entry
Czech Republic
ContactUsLinkComponent
 
 
 
 
Sodium dodecyl sulphate (SDS) ≥98%, GPR RECTAPUR®
 
undefined
 
 
 
Sodium dodecyl sulphate (SDS) ≥98%, GPR RECTAPUR®
 

 

  • Assay (calculated on anhydrous):
    Min. 98 %
  • Cl (Chloride):
    Max. 0.1 %
  • PO4 (Phosphate):
    Max. 10 ppm

 

Frequently Bought Together