0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
1,6-Hexanedithiol
1,6-Hexanedithiol ≥97%
1,6-Hexanedithiol ≥97%
:
Thermo Fisher Scientific
1,6-Hexanedithiol ≥97%
:
Thermo Fisher Scientific
:
1191-43-1
:
:
C₆H₁₄S₂
:
MFCD00004910
:
242…243 °C
:
–21 °C
:
0.990
:
90 °C (194 °F)
:
Ambient
:
214-735-1
:
9,III
:
150.31 g/mol
:
3334
msds_ALFAL06686
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Methanol ≥99.9% (by GC), HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Chloroform 99.0-99.4% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
Chloroform, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals