
1,4-Butanedithiol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1191-08-8
:
Formula:C₄H₁₀S₂
MDL Number:MFCD00004906
:122.26 g/mol
Boiling Pt:105 °C (1013 hPa)
Melting Pt:–53 °C
Density:1.0621 g/cm³ (0 °C)
EINECS:214-728-3
ADR:9,
:M-Clarity™ quality level = MQ300
UN:3334
:GHS07
Storage Temperature:Ambient
Frequently Bought Together