

1,2-Ethanedithiol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:540-63-6
:
Formula:C₂H₆S₂
MDL Number:MFCD00004892
:94.2 g/mol
Merck Index:13,03760
Boiling Pt:146…149 °C (1013 hPa)
Melting Pt:–41 °C
Density:1.374 g/cm³ (20 °C)
EINECS:208-752-3
UN:3071
ADR:6.1,II
:M-Clarity™ quality level = MQ200
Flash Pt:44 °C
:GHS02
:GHS06
Storage Temperature:Ambient
Frequently Bought Together