Order Entry
Czech Republic
ContactUsLinkComponent
1-Hexadecanethiol for synthesis, Sigma-Aldrich®
1-Hexadecanethiol for synthesis, Sigma-Aldrich®
  8.22063.0250
 :  Merck
 :  
1-Hexadecanethiol for synthesis, Sigma-Aldrich®
  8.22063.0250
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590

 

 

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    18 - 20 °C
  • Molar Mass:
    258.5 g/mol
  • Boiling Point:
    185 - 190 °C (9 hPa)
  • Vapor Pressure:
     - 0.1 hPa (20 °C)
  • Flash Point:
    102 °C
  • Refractive Index:
    1.462 (20 °C, 589 nm)
  • Density:
    0.85 g/cm3 (20 °C)
  • Ignition Point:
    260 °C

 

 

 :
 :258.51 g/mol
 :335 °C (1013 hPa)
 :18…20 °C
 :0.8556 g/cm³ (20 °C)
 :99 °C
 :Ambient
 :220-846-6
 :3334
 :9,
 :M-Clarity™ quality level = MQ200
 :2917-26-2
 :C₁₆H₃₄S
 :MFCD00011677