


Ethanethiol for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:75-08-1
:
Formula:C₂H₆S
MDL Number:MFCD00004887
Merck Index:12,03771
Boiling Pt:35 °C (1013 hPa)
Melting Pt:–148 °C
Density:0.846 g/cm³ (20 °C)
Flash Pt:–45 °C
EINECS:200-837-3
UN:2363
ADR:3,I
:M-Clarity™ quality level = MQ200
:62.14 g/mol
:GHS02
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Solubility:6.8 g/l (20 °C)
- Melting Point:-148 °C
- Molar Mass:62.14 g/mol
- Boiling Point:35 °C (1013 hPa)
- Vapor Pressure:576 hPa (20 °C)
- Flash Point:-45 °C
- Refractive Index:1.4306 (20 °C, 589 nm)
- Explosion Limit:2.8 - 18 %(V)
- Density:0.84 g/cm3 (20 °C)
- Ignition Point:295 °C
Specification Test Results
Frequently Bought Together