Order Entry
Czech Republic
ContactUsLinkComponent
1-Dodecanethiol for synthesis, Sigma-Aldrich®
Corrosive
Irritant
Environment
1-Dodecanethiol for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Corrosive
Irritant
Environment
1-Dodecanethiol for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
     - 1 g/l (22 °C)
  • Melting Point:
    -7 °C
  • Molar Mass:
    202.39 g/mol
  • Boiling Point:
    275 °C (1013 hPa)
  • Vapor Pressure:
    0.002 hPa (20 °C)
  • Flash Point:
    127 °C
  • Refractive Index:
    1.4587 (20 °C, 589 nm)
  • Density:
    0.845 g/cm3 (20 °C)
  • Ignition Point:
    230 °C
  • Synonyms:
    Lauryl mercaptan, Dodecyl mercaptan
  • Hill Formula:
    C12H26S
  • Chemical Formula:
    CH3(CH2)11SH

Specification Test Results

Related Information

Frequently Bought Together