Order Entry
Czech Republic
ContactUsLinkComponent
1,2,6-Hexanetriol 96%
1,2,6-Hexanetriol 96%
Katalogové číslo L06032.14
CAS Number:  
1,2,6-Hexanetriol 96%
Katalogové číslo L06032.14
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :134.18 g/mol
Boiling Pt:166…168 °C (2 mmHg)
Melting Pt:–20 °C
Density:1.106
Flash Pt:191 °C (375 °F)
Storage Temperature:Ambient
EINECS:203-424-6
CAS Number:106-69-4
Formula:C₆H₁₄O₃
MDL Number:MFCD00002976