Order Entry
Czech Republic
ContactUsLinkComponent
1,5-Pentanediol for synthesis, Sigma-Aldrich®
1,5-Pentanediol for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
1,5-Pentanediol for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    -16 °C
  • Molar Mass:
    104.15 g/mol
  • Boiling Point:
    240 - 242 °C (1013 hPa)
  • Vapor Pressure:
    0.01 hPa (20 °C)
  • Flash Point:
    136 °C
  • Refractive Index:
    1.4494 (20 °C, 589 nm)
  • Explosion Limit:
    1.3 - 13.1 %(V)
  • Density:
    0.99 g/cm3 (20 °C)
  • pH:
    7.5 (H2O)
  • Ignition Point:
    330 °C

Specification Test Results

Related Information

Frequently Bought Together