Order Entry
Czech Republic
ContactUsLinkComponent
Ethylene glycol ≥98%, TECHNICAL
Irritant
Ethylene glycol ≥98%, TECHNICAL
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates
undefined
Irritant
Ethylene glycol ≥98%, TECHNICAL
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates
Limited Time Sale
Select items below are currently on sale

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 98 %
  • Appearance of solution (50 % V/V; water):
    Passes test

Specification Test Results

Frequently Bought Together