Order Entry
Czech Republic
ContactUsLinkComponent
Ethanol absolute (taxed) ≥99.8% (by GC)
Ethanol absolute (taxed) ≥99.8% (by GC)
Certificates
Ethanol absolute (taxed) ≥99.8% (by GC)

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay (GC):
    Min. 99.8 Vol.%
  • Density (D 20/20):
    0.790 - 0.791
  • Non-volatile matter:
    Max. 0.01 %
  • Water (Karl Fischer):
    Max. 0.2 %
  • Free acid (as CH₃COOH):
    Max. 0.001 %
  • Free alkali (as NH₃):
    Max. 0.0005 %
  • Aldehydes (as CH₃CHO):
    Max. 0.005 %
  • Methanol (GC):
    Max. 0.1 %
  • Autoignition Temperature:
    683 °F
  • Explosion Limit:
    19 %, 60 °F

Specification Test Results

Related Information

Frequently Bought Together