(±)-3-Nonanol for synthesis, Sigma-Aldrich®
Catalog # 8.18784.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 ml
- Seq:2315
Specifications
Specification Test Results
- Molar Mass:144.25 g/mol
- Boiling Point:91 - 92 °C (39 hPa)
- Flash Point:96 °C
- Refractive Index:1.4289 (20 °C, 589 nm)
- Density:0.82 g/cm3 (20 °C)
- Synonyms:Ethyl hexyl carbinol
- Hill Formula:C9H20O
- Chemical Formula:CH3(CH2)5CH(OH)CH2CH3
Specification Test Results
About this item
:
:144.26 g/mol
Boiling Pt:195 °C (1013 hPa)
Melting Pt:–23…–22 °C
Density:0.8211 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:210-850-6
CAS Number:624-51-1
Formula:C₉H₂₀O
MDL Number:MFCD00014411