(±)-3-Hexanol
Catalog # EHERC14196900
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- Seq:2070
Specifications
About this item
:
Boiling Pt:135 °C (1013 hPa)
Melting Pt:–70 °C
Density:0.819 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:3,III
:102.18 g/mol
EINECS:210-790-0
UN:2282
CAS Number:623-37-0
Formula:C₆H₁₄O
MDL Number:MFCD00004582