

2-Propanol-[D₈] 99+ atom % D for NMR spectroscopy
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:22739-76-0
:
Formula:D₃CCD(OD)CD₃
MDL Number:MFCD00044341
:68.03 g/mol
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–89.5 °C
Density:0.89 g/cm³
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:245-189-2
UN:1219
ADR:3,II
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.05 %
- D-Enrichment:≥99.0 %
- GC:≥98.5 % (sum of isomers)
Specification Test Results
Frequently Bought Together