Order Entry
Czech Republic
ContactUsLinkComponent
1-Tetradecanol 99%
Irritant
Environment
1-Tetradecanol 99%
Certificates
undefined
Irritant
Environment
1-Tetradecanol 99%

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals or flakes or pellets and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    37 to 41 °C
  • GC:
    ≥98.5 %
  • Titration with HClO4:
    97.5 to 102.5 % (on dry substance)

Specification Test Results

Related Information

Frequently Bought Together