
1-Octanol ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:111-87-5
:
Formula:H₃C(CH₂)₆CH₂OH
MDL Number:MFCD00002988
:130.23 g/mol
Merck Index:14,6751
Boiling Pt:193…195 °C
Melting Pt:–16 °C
Density:0.825
Flash Pt:81 °C (177 °F)
EINECS:203-917-6
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together