1-Butanol, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:71-36-3
:
Formula:H₃C(CH₂)₂CH₂OH
MDL Number:MFCD00002964
:74.12 g/mol
Merck Index:13,01539
Boiling Pt:117.6 °C (1013 hPa)
Melting Pt:–89 °C
Density:0.812 g/cm³ (20 °C)
Flash Pt:30 °C
Storage Temperature:Ambient
EINECS:200-751-6
UN:1120
ADR:3,III
REACH:01-2119484630-38
:GHS02
:GHS05
:GHS07
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
Specifications
Specification Test Results
- Assay (by GC):min. 99.4 %
- Aldehydes:passes test
- Butyl Ether (by GC):max. 0.2 %
- Carbonyl Compounds (as butyraldehyde):max. 0.01 %
- Color (APHA):max. 10
- Residue after Evaporation:max. 0.001 %
- Titrable Acid (meq/g):max. 0.0008
- Water (H2O):max. 0.1 %
- Aluminium (Al):max. 0.5
- Boron (B):max. 0.02
- Barium (Ba):max. 0.1
- Calcium (Ca):max. 0.5
- Cadmium (Cd):max. 0.05
- Cobalt (Co):max. 0.02
- Chromium (Cr):max. 0.02
- Copper (Cu):max. 0.02
- Iron (Fe):max. 0.1
- Magnesium (Mg):max. 0.1
- Manganese (Mn):max. 0.02
- Nickel (Ni):max. 0.02
- Lead (Pb):max. 0.1
- Tin (Sn):max. 0.1
- Zinc (Zn):max. 0.1
Specification Test Results
Frequently Bought Together