

Formaldehyde diethyl acetal 99% stabilised, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:462-95-3
:
Formula:C₅H₁₂O₂
MDL Number:MFCD00009246
:104.15 g/mol
Boiling Pt:88 °C (1013 hPa)
Melting Pt:–66.5 °C
Density:0.831 g/cm³
Flash Pt:–5 °C
Storage Temperature:Ambient
EINECS:207-330-6
UN:2373
ADR:3,II
:GHS02
:GHS06
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3720 to 1.3760 (20°C, 589 nm)
- GC:≥98.5 %
- Stabilizer:50 to 150 ppm (BHT)
- Peroxides:≤10 ppm
Specification Test Results
Frequently Bought Together