Order Entry
Czech Republic
ContactUsLinkComponent
Diethyl ether, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
Irritant
Flammable
Diethyl ether, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
Certificates
undefined
Irritant
Flammable
Diethyl ether, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®

Specification Test Results

  • Appearance:
    clear colorless liquid
  • GC:
    ≥99.4%
  • Water:
    ≤0.005% (Coulometric)
  • Acidity:
    ≤0.0002 meq/g
  • Peroxides:
    ≤0.002% (as H2O2)

Specification Test Results

Frequently Bought Together