Isopropyl isocyanate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1795-48-8
:
Formula:C₄H₇NO
MDL Number:MFCD00002037
:85.11 g/mol
Boiling Pt:74 °C (1013 hPa)
Density:0.8669 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:217-276-5
UN:2483
ADR:3,I
:M-Clarity™ quality level = MQ200
Frequently Bought Together