Order Entry
Czech Republic
ContactUsLinkComponent
Iodomethane ≥99%, GPR RECTAPUR®
Health
Toxic
Iodomethane ≥99%, GPR RECTAPUR®
Catalog # 25596.154
Supplier:  VWR Chemicals
CAS Number:  
undefined
Health
Toxic
Iodomethane ≥99%, GPR RECTAPUR®
Catalog # 25596.154
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • IR Spectrum:
    Passes test
  • Density (20/4):
    2.260 - 2.290

Specification Test Results

About this item

 :
 :141.94 g/mol
Merck Index:12,06161
Boiling Pt:42.4 °C (1013 hPa)
Melting Pt:–66 °C
Density:2.279 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:200-819-5
UN:2644
ADR:6.1,I
 :GHS08
 :GHS06
CAS Number:74-88-4
Formula:CH₃I
MDL Number:MFCD00001073