


Iodomethane stabilised for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:74-88-4
:
Formula:CH₃I
MDL Number:MFCD00001073
:141.94 g/mol
Merck Index:12,06161
Boiling Pt:42.4 °C (1013 hPa)
Melting Pt:–66 °C
Density:2.279 g/cm³ (20 °C)
EINECS:200-819-5
UN:2644
ADR:6.1,I
:M-Clarity™ quality level = MQ200
:Stabilised with silver
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
Frequently Bought Together