Order Entry
Czech Republic
ContactUsLinkComponent
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10-Heneicosafluoro-12-iodododecane, Sigma-Aldrich®
Irritant
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10-Heneicosafluoro-12-iodododecane, Sigma-Aldrich®
Catalog # SIAL91987-2.5G-F
Supplier:  Merck
CAS Number:  
Irritant
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10-Heneicosafluoro-12-iodododecane, Sigma-Aldrich®
Catalog # SIAL91987-2.5G-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2,5 g
  • Seq:
    3122

Specifications

About this item

 :
 :674.03 g/mol
EINECS:218-054-0
 :GHS07
Storage Temperature:Ambient
CAS Number:2043-54-1
Formula:C₁₂H₄F₂₁I
MDL Number:MFCD00039412