Order Entry
Czech Republic
ContactUsLinkComponent
1-Chloro-2-propanol ≥70%, Sigma-Aldrich®
Flammable
Toxic
1-Chloro-2-propanol ≥70%, Sigma-Aldrich®
Catalog # SIAL292087-25G
Supplier:  Merck
CAS Number:  
Flammable
Toxic
1-Chloro-2-propanol ≥70%, Sigma-Aldrich®
Catalog # SIAL292087-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :94.54 g/mol
Merck Index:12,08036
Boiling Pt:126 °C (1013 hPa)
Density:1.1302 g/cm³ (0 °C)
Storage Temperature:Ambient
EINECS:204-819-6
UN:2929
ADR:6.1,II
 :M-Clarity™ quality level = MQ200
 :GHS02
 :GHS06
CAS Number:127-00-4
Formula:C₃H₇ClO
MDL Number:MFCD00004530