

1,3-Dichloro-2-propanol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:96-23-1
:
Formula:ClH₂CCHOHCH₂Cl
MDL Number:MFCD00000951
:128.99 g/mol
Merck Index:14,3074
Boiling Pt:174…176 °C
Melting Pt:–4 °C
Density:1.366
Flash Pt:85 °C (185 °F)
Storage Temperature:Ambient
EINECS:202-491-9
UN:2750
ADR:6.1,II
:GHS06
:GHS08
Related Information
Frequently Bought Together