Chloroform ≥99.8% stabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane
CAS Number:67-66-3
:
Formula:CHCl₃
MDL Number:MFCD00000826
Boiling Pt:61.7 °C (1013 hPa)
Density:1.4832 g/cm³ (20 °C)
:20 ppm Stabilised with 2-methyl-2-butene (iso-amylene)
Molecular Weight:119.38 g/mol
Melting Pt:–63 °C
EINECS:200-663-8
UN:1888
ADR:6.1 III
Merck Index:13,02160
:Filtered through a 0.2 µm filter, packaged under nitrogen.
REACH:01-2119486657-20
:GHS08
:GHS06
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (GC):Min. 99.80 %
- Water:Max. 0.0500 %
- Non-volatile residue:Max. 0.001 %
- Acidity:Max. 0.0005 meq/g
- Alkalinity:Max. 0.0002 meq/g
- Transmittance (250 nm):Min. 50.0 %
- Transmittance (260 nm):Min. 84.00 %
- Transmittance (280 nm):Min. 95.0 %
- Transmittance (300 nm):Min. 98.0 %
- Conforms to BDH 15283:Passes test
Specification Test Results
Related Information
Frequently Bought Together