Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Chloroform ≥98% stabilised, TECHNICAL

Chloroform ≥98% stabilised, TECHNICAL

Health
Toxic
Supplier:   VWR Chemicals
Chloroform ≥98% stabilised, TECHNICAL
Supplier:  VWR Chemicals
Certificates
Chloroform ≥98% stabilised, TECHNICAL

3 Options Available Below

Product Details & Documents
Synonyms:  Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane
  • CAS Number:67-66-3
  • Formula:CHCl₃
  • MDL Number:MFCD00000826
  • Molecular Weight:119.38 g/mol
  • Merck Index:13,02160
  • Boiling Pt:61.7 °C (1013 hPa)
  • Melting Pt:–63 °C
  • Density:1.4832 g/cm³ (20 °C)
  • EINECS:200-663-8
  • ADR:6.1,III
  •  :Stabilised with ethanol 0.6 %
  • Storage Temperature:Ambient
  • UN:1888
  • REACH:01-2119486657-20
Specifications for Product Family

Specification Test Results

  • Assay:
    Min. 98 %
  • Density (20/4):
    1.473 - 1.489
  • Ethanol:
    0.1 - 1.0 %
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.