Chloroform, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
: Avantor
: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane
:67-66-3
:
:CHCl₃
:MFCD00000826
:61.7
:–63 °C
:1.4832 g/cm³ (20 °C)
:119.38 g/mol
:Ambient
:200-663-8
:6.1 III
:13,02160
:1888
:05-2114614906-45
:GHS06
:GHS08
- Environmentally Preferable:
- Acetone and Aldehyde (as (CH3)2CO):max. 0.005 %
- Acid and Chloride:passes test
- Density (g/ml) at 25°C:1.467 - 1.474
- Free Chlorine:passes test
- Preservative (C2H5OH):about 0.75%
- Substances Darkened by H2SO4:passes test
- Aluminium (Al):max. 0.5
- Boron (B):max. 0.02
- Barium (Ba):max. 0.1
- Calcium (Ca):max. 0.5
- Cadmium (Cd):max. 0.05
- Cobalt (Co):max. 0.02
- Chromium (Cr):max. 0.02
- Copper (Cu):max. 0.02
- Iron (Fe):max. 0.1
- Magnesium (Mg):max. 0.1
- Manganese (Mn):max. 0.02
- Nickel (Ni):max. 0.02
- Lead (Pb):max. 0.1
- Tin (Sn):max. 0.1
- Zinc (Zn):max. 0.1
- Assay (by GC):min. 99 %
- Color (APHA):max. 10
- Residue after Evaporation:max. 0.001 %
Frequently Bought Together