


1,2-Dichloroethane 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:107-06-2
:
Formula:ClH₂CCH₂Cl
MDL Number:MFCD00000963
:98.96 g/mol
Merck Index:13,03831
Boiling Pt:81…85 °C (760 mmHg)
Melting Pt:–35 °C
Density:1.25 g/cm³
Flash Pt:13 °C
Storage Temperature:Ambient
EINECS:203-458-1
UN:1184
ADR:3,II
:GHS06
:GHS08
:GHS02
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.03 %
- GC:≥99.0 %
- UV:at 230 nm A: ≤1
- Color scale:≤10 APHA
- Residue after evaporation:≤0.002 %
Specification Test Results
Frequently Bought Together