Order Entry
Czech Republic
ContactUsLinkComponent
 
 
Barium acetate, EMSURE® ACS for analysis, Supelco®
  1.01704.0500
 :  Merck
 :  
 
Barium acetate, EMSURE® ACS for analysis, Supelco®
  1.01704.0500
 :  Merck
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645

 

 

  • Solubility:
    720 g/l (20 °C)
  • Melting Point:
    450 °C
  • Molar Mass:
    255.42 g/mol
  • Bulk Density:
    1200 kg/m3
  • Density:
    2.47 g/cm3 (20 °C)
  • pH:
    6.5 - 8.5 (50 g/l, H2O, 20 °C)
  • Hazard Class:
    6.1
  • Hazard Class Description:
    Toxic Materials

 

 

 :
 :255.42 g/mol
 :13,00968
 :∼450 °C
 :2.468 g/cm³ (20 °C)
 :Ambient
 :208-849-0
 :1564
 :6.1,III
 :M-Clarity™ quality level = MQ300
 :GHS07
 :543-80-6
 :(H₃CCOO)₂Ba
 :MFCD00012447