
Myristic acid for synthesis, Sigma-Aldrich®
: Merck
:544-63-8
:
:C₁₄H₂₈O₂
:MFCD00002744
:228.38 g/mol
:12,06416
:319 °C (1013 hPa)
:51…54 °C
:0.8622 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:>165 °C
:208-875-2
:GHS07
:Ambient
- Solubility:1.07 mg/l (25 °C)
- Melting Point:52 - 56 °C
- Molar Mass:228.37 g/mol
- Boiling Point:326 °C (1013 hPa)
- Vapor Pressure:- 0.01 hPa (20 °C)
- Flash Point:165 °C
- Density:0.8622 g/cm3 (54 °C)
- Synonyms:Tetradecanoic acid
- Hill Formula:C14H28O2
- Chemical Formula:CH3(CH2)12COOH
Frequently Bought Together