


Propionic acid ≥99.5% (by GC) eluent additive, for LC-MS, Fluka™
49916-50ML
:
- Pack type:Plastic bottle
- Pk:50 mL
- Seq:2370
- Refractive Index:n20/D 1.386 (lit.)
- Identity (NMR):complying
- Purity (GC):Min. 99.5 %
- Assay (alkalimetric):99.5 - 100.5 %
- Refractive index (n 20/D):1.385 - 1.387
- Water (Karl Fischer):Max. 0.1 %
- Aluminium (Al):Max. 0.5 ppm
- Barium (Ba):Max. 0.1 ppm
- Bismuth (Bi):Max. 0.1 ppm
- Calcium (Ca):Max. 0.5 ppm
- Cadmium (Cd):Max. 0.05 ppm
- Cobalt (Co):Max. 0.02 ppm
- Chromium (Cr):Max. 0.1 ppm
- Copper (Cu):Max. 0.02 ppm
- Iron (Fe):Max. 0.5 ppm
- Potassium (K):Max. 0.5 ppm
- Lithium (Li):Max. 0.1 ppm
- Magnesium (Mg):Max. 0.1 ppm
- Manganese (Mn):Max. 0.02 ppm
- Molybdenum (Mo):Max. 0.1 ppm
- Sodium (Na):Max. 1 ppm
- Nickel (Ni):Max. 0.1 ppm
- Lead (Pb):Max. 0.1 ppm
- Strontium (Sr):Max. 0.1 ppm
- Zinc (Zn):Max. 0.1 ppm
- Suitability for the LC-MS:complying
:
:74.08 g/mol
:13,07917
:141 °C (1.013 hPa)
:-21 °C
:–21 °C
:0.99 g/cm³ (20 °C)
:12.0 %(V)
:Ambient
:201-176-3
:3463
:8,II
:01-2119486971-24
:GHS02
:GHS05
:GHS07
:79-09-4
:C₃H₆O₂
:MFCD00002756