


Formic acid 98.0-100%, Puriss., meets analytical specifications of DAC, FCC, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:64-18-6
:
Formula:HCO₂H
MDL Number:MFCD00003297
:46.03 g/mol
Boiling Pt:100.4 °C (1.013 hPa)
Melting Pt:4 °C
Density:1.22 g/cm³ (20 °C)
Flash Pt:49.5 °C
Storage Temperature:Ambient
EINECS:200-579-1
UN:1779
ADR:8,II
:GHS02
:GHS06
:GHS05
Specification Test Results
- Appearance:complying
- Identity:complying
- Assay:98.0 - 100.5 %
- Density (D 20/20):1.217 - 1.223
- Non-volatile matter:Max. 0.005 %
- Iron (Fe):Max. 0.001 %
- Heavy metals (as Pb):Max. 10 ppm
- Chloride (Cl):Max. 10 ppm
- Sulfate (SO₄):Max. 40 ppm
- Sulfite (SO₃):complying
- Acetic acid (GC):Max. 0.05 %
- Solubility in water:complying
- Residual solvents:complying
- Chromatographic purity:complying
- Refractive Index:n20/D 1.370 (lit.)
Specification Test Results
Frequently Bought Together