Acetic acid glacial 99.8-100.0%, ARISTAR® for trace analysis
Catalog # 450013Y
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle, Safebreak®
- Pk:500 mL
- Seq:2725
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:99.8 - 100.0 %
- Colour:Max. 10 APHA
- Acetaldehyde:Max. 2 ppm
- Acetone:Max. 1 ppm
- Acetic anhydride:Max. 100 ppm
- Freezing point:Min. 16.3 °C
- Cl (Chloride):Max. 0.1 ppm
- PO4 (Phosphate):Max. 0.1 ppm
- SO4 (Sulphate):Max. 0.5 ppm
- Ag (Silver):Max. 0.010 ppm
- Al (Aluminium):Max. 0.005 ppm
- As (Arsenic):Max. 0.003 ppm
- Au (Gold):Max. 0.020 ppm
- B (Boron):Max. 0.02 ppm
- Ba (Barium):Max. 0.005 ppm
- Be (Beryllium):Max. 0.010 ppm
- Bi (Bismuth):Max. 0.050 ppm
- Ca (Calcium):Max. 0.200 ppm
- Cd (Cadmium):Max. 0.003 ppm
- Cr (Chromium):Max. 0.010 ppm
- Co (Cobalt):Max. 0.005 ppm
- Cu (Copper):Max. 0.003 ppm
- Fe (Iron):Max. 0.050 ppm
- Ga (Gallium):Max. 0.010 ppm
- Ge (Germanium):Max. 0.020 ppm
- Hg (Mercury):Max. 0.002 ppm
- In (Indium):Max. 0.010 ppm
- K (Potassium):Max. 0.050 ppm
- Li (Lithium):Max. 0.010 ppm
- Mg (Magnesium):Max. 0.010 ppm
- Mn (Manganese):Max. 0.005 ppm
- Mo (Molybdenum):Max. 0.005 ppm
- Na (Sodium):Max. 0.200 ppm
- Ni (Nickel):Max. 0.005 ppm
- Pb (Lead):Max. 0.005 ppm
- Pt (Platinum):Max. 0.050 ppm
- Sn (Tin):Max. 0.050 ppm
- Sr (Strontium):Max. 0.005 ppm
- Ti (Titanium):Max. 0.050 ppm
- Tl (Thallium):Max. 0.050 ppm
- V (Vanadium):Max. 0.010 ppm
- Zn (Zinc):Max. 0.050 ppm
- Zr (Zirconium):Max. 0.050 ppm
- Substances reducing K2Cr2O7:Passes test
- Substances reducing KMnO4:Passes test
- Dilution test:Passes test
- Non-volatile matter:Max. 5 ppm
Specification Test Results
About this item
Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid
:
:60.05 g/mol
Boiling Pt:118 °C (1013 hPa)
Melting Pt:17 °C
Density:1.05 g/cm³ (20 °C)
Flash Pt:38.5 °C
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
ADR:8 (3),II
REACH:01-2119475328-30
:GHS02
:GHS05
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152