Order Entry
Czech Republic
ContactUsLinkComponent
Ethyl acetate, LiChrosolv® for HPLC, Supelco®
Flammable
Irritant
Ethyl acetate, LiChrosolv® for HPLC, Supelco®
Supplier:  Merck
   Sustainable Options Available
Certificates
undefined
Flammable
Irritant
Ethyl acetate, LiChrosolv® for HPLC, Supelco®
Supplier:  Merck
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • Solubility:
    85.3 g/l (20 °C)
  • Melting Point:
    -83 °C
  • Molar Mass:
    88.11 g/mol
  • Boiling Point:
    77 °C (1013 hPa)
  • Vapor Pressure:
    97 hPa (20 °C)
  • Flash Point:
    -4 °C
  • Refractive Index:
    1.37
  • Explosion Limit:
    2.1 - 11.5 %(V)
  • Density:
    0.90 g/cm3 (20 °C)
  • Ignition Point:
    460 °C
  • Synonyms:
    Ethyl ethanoate, Acetic ester, Ethyl ester
  • Hill Formula:
    C4H8O2
  • Chemical Formula:
    CH3COOC2H5

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together