

Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
:141-78-6
:
:CH₃COOC₂H₅
:MFCD00009171
:88.11 g/mol
:13,03792
:77 °C (1.013 hPa)
:–84 °C
:0.901 g/cm³
:11.5 %(V)
:Ambient
:205-500-4
:1173
:3,II
:01-2119475103-46
:GHS07
:GHS02
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
- Assay (GC):Min. 99.5 %
- Boiling range:76 - 78 °C
- Density (D 20/4):0.899 - 0.901
- Refractive index (n 20/D):1.3710 - 1.3730
- Non-volatile matter:Max. 0.001 %
- Water (Karl Fischer):Max. 0.05 %
- Free acid (as CH₃COOH):Max. 0.0045 %
- Aluminium (Al):Max. 0.00005 %
- Boron (B):Max. 0.000002 %
- Barium (Ba):Max. 0.00001 %
- Bismuth (Bi):Max. 0.00001 %
- Calcium (Ca):Max. 0.00005 %
- Cadmium (Cd):Max. 0.000005 %
- Cobalt (Co):Max. 0.000002 %
- Chromium (Cr):Max. 0.000002 %
- Copper (Cu):Max. 0.000002 %
- Iron (Fe):Max. 0.00001 %
- Potassium (K):Max. 0.00005 %
- Lithium (Li):Max. 0.00001 %
- Magnesium (Mg):Max. 0.00001 %
- Molybdenum (Mo):Max. 0.00001 %
- Manganese (Mn):Max. 0.000002 %
- Sodium (Na):Max. 0.00005 %
- Nickel (Ni):Max. 0.000002 %
- Lead (Pb):Max. 0.00001 %
- Tin (Sn):Max. 0.00001 %
- Strontium (Sr):Max. 0.00001 %
- Zinc (Zn):Max. 0.00001 %
- methyl acetate:Max. 0.1 %
- Ethanol (GC):Max. 0.1 %
- Isobutyl alcohol (GC):Max. 0.1 %
- Methanol (GC):Max. 0.1 %
- Reaction against H₂SO₄:complying
- APHA:Max. 10
Frequently Bought Together