

3,4-Hexanedione 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4437-51-8
:
Formula:C₆H₁₀O₂
MDL Number:MFCD00010237
:114.14 g/mol
Boiling Pt:129 °C (1013 hPa)
Melting Pt:–10 °C
Density:0.941 g/cm³
Flash Pt:29 °C
Storage Temperature:Ambient
EINECS:224-651-7
UN:1224
ADR:3,III
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4090 to 1.4150 (20°C, 589 nm)
- GC:≥95 %
- Specific gravity:(25°C) 0.93 to 0.95
Specification Test Results
Frequently Bought Together