Order Entry
Czech Republic
ContactUsLinkComponent
Acetylacetone ≥99%, TECHNICAL
Flammable
Irritant
Acetylacetone ≥99%, TECHNICAL
Catalog # 20081.298
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Irritant
Acetylacetone ≥99%, TECHNICAL
Catalog # 20081.298
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Refractive index (20°C):
    1.450 - 1.452

Specification Test Results

About this item

 :
 :100.12 g/mol
Merck Index:13,00083
Boiling Pt:140.4 °C (1013 hPa)
Melting Pt:–23 °C
Density:0.96945 g/cm³ (20 °C)
Flash Pt:30 °C
Storage Temperature:Ambient
EINECS:204-634-0
UN:2310
ADR:3 (6.1),III
 :GHS02
 :GHS07
CAS Number:123-54-6
Formula:CH₃COCH₂COCH₃
MDL Number:MFCD00008787