VWR® Rota-Rack Rotatable Racks for Test Tubes
Supplier: VWR Collection
Certificates
About this item
PP.
- Excellent thermal resistance, autoclavable
- Type A: With one side each for 2×50 ml tubes, 8×15 ml tubes, 6×20 mm tubes and 10 tubes with 10 to 13 mm diameter
- Type B: With one side each for 6×15 ml tubes, 9×¹,⁵/₂,₀ ml microcentrifuge tubes, 12×⁰,⁵/₀,₆ ml microcentrifuge tubes and 32×0,2 ml PCR tubes or four strips with eight PCR tubes
Racks with four independently-rotating modules coloured purple, blue, green and yellow for simultaneous loading with test tubes, culture tubes, centrifuge tubes or microcentrifuge tubes of different sizes.
Frequently Bought Together