ContactUsLinkComponent
Petri dish racks, Nalgene®
Petri dish racks, Nalgene®
Catalog # NALG5922-0024
   Low Manufacturing Impact
undefined
Petri dish racks, Nalgene®
Catalog # NALG5922-0024
   Low Manufacturing Impact
Specifications
  • Description:
    Petri dish rack
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    NAR Certificate of Retirement
  • Pk:
    1
Specifications
About this item
Petri dish rack with acrylic frame and PC posts.
  • Lightweight
  • Easy to load
  • Provides safe transport
Break-resistant PC posts. Supplied unassembled. Holds up to twenty-four 100 mm dishes.