Order Entry
Czech Republic
ContactUsLinkComponent
2-Tridecanone
2-Tridecanone
Catalog # EHERC17818500
Supplier:  EHRENSTORFER
CAS Number:  
2-Tridecanone
Catalog # EHERC17818500
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :198.35 g/mol
Boiling Pt:263 °C (1013 hPa)
Melting Pt:25…27 °C
Density:0.82 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:209-784-0
CAS Number:593-08-8
Formula:C₁₃H₂₆O
MDL Number:MFCD00008968