

2-Pentanone for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:107-87-9
:
Formula:C₅H₁₀O
MDL Number:MFCD00009400
:86.13 g/mol
Merck Index:12,06193
Boiling Pt:100 °C (1013 hPa)
Melting Pt:–78 °C
Density:0.809 g/cm³ (20 °C)
EINECS:203-528-1
UN:1249
ADR:3,II
:M-Clarity™ quality level = MQ200
Flash Pt:7 °C
:GHS02
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:43 g/l (25 °C)
- Melting Point:-78 °C
- Molar Mass:86.13 g/mol
- Boiling Point:101 - 103 °C (1013 hPa)
- Vapor Pressure:37 hPa (20 °C)
- Flash Point:7 °C
- Refractive Index:1.3895 (20 °C, 589 nm)
- Explosion Limit:1.5 - 8.2 %(V)
- Density:0.81 g/cm3 (20 °C)
- Ignition Point:449 °C
Specification Test Results
Frequently Bought Together