
2-Pentanone
Catalog # EHERC15981500
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- Seq:2070
Specifications
About this item
:
:86.13 g/mol
Merck Index:12,06193
Boiling Pt:100 °C (1013 hPa)
Melting Pt:–78 °C
Density:0.809 g/cm³ (20 °C)
Flash Pt:7 °C
Storage Temperature:Ambient
EINECS:203-528-1
UN:1249
ADR:3,II
:GHS02
CAS Number:107-87-9
Formula:C₅H₁₀O
MDL Number:MFCD00009400