

N,O-Bis(trimethylsilyl)trifluoroacetamide
Catalog # HYPETS-38830
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:10 Ampoul
- Seq:0510
Specifications
About this item
:
:257.4 g/mol
Boiling Pt:145 °C (1013 hPa)
Melting Pt:–41 °C
Density:0.96 g/cm³ (20 °C)
Flash Pt:34 °C
EINECS:247-103-9
UN:2924
ADR:3,II
:GHS02
:GHS07
CAS Number:25561-30-2
Formula:C₈H₁₈F₃NOSi₂
MDL Number:MFCD00008269