Ethyl acrylate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:140-88-5
:
Formula:C₅H₈O₂
MDL Number:MFCD00009188
:100.12 g/mol
ADR:3,II
:M-Clarity™ quality level = MQ200
EINECS:205-438-8
UN:1917
Frequently Bought Together