



Methylmagnesium bromide 1 M in THF, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75-16-1
:
Formula:CH₃BrMg
MDL Number:MFCD00000041
:119.24 g/mol
Density:1 g/cm³
Flash Pt:–21 °C
Storage Temperature:Ambient
EINECS:200-844-1
UN:3399
ADR:4.3,I
:GHS05
:GHS02
:GHS07
:GHS08
Specification Test Results
- Appearance:Clear to turbid grey to dark purple to brown-black Solution (May develop some turbidity or precipitate)
- Titration Grignard:0.95 to 1.20 M
Specification Test Results
Related Information
Frequently Bought Together