

Methanol ≥99.7% (by GC), Puriss., meets analytical specification of Ph.Eur., Riedel-de Haën™
:67-56-1
:
:CH₃OH
:MFCD00004595
:32.04 g/mol
:13,05984
:64…65 °C (1.013 hPa)
:–98 °C
:0.79 g/cm³ (20 °C)
:11 °C
:Ambient
:200-659-6
:1230
:3,II
:01-2119433307-44
:GHS08
:GHS02
- Assay (GC):Min. 99.7 %
- Boiling range:64 - 65 °C
- Density (D 20/20):0.791 - 0.793
- Refractive index (n 20/D):1.328 - 1.330
- Non-volatile matter:Max. 10 ppm
- Water (Karl Fischer):Max. 0.10 %
- Acidity or alkalinity:complying
- Free acid (as HCOOH):Max. 0.003 %
- Free alkali (as NH₃):Max. 0.0003 %
- Iron (Fe):Max. 0.0001 %
- Identity (IR):complying
- Chloride (Cl):Max. 0.0001 %
- Sulfate (SO₄):Max. 0.0005 %
- Reaction against H₂SO₄:complying
- Reducing impurities:complying
- Aldehydes, Ketones (as CH₃CocH₃):Max. 0.003 %
- Acetone (GC):Max. 0.001 %
- Benzene (GC):Max. 2 ppm
- UV-absorption:complying
- Impurity A (GC):complying
- Related substances (GC):complying
- Appearance:complying
- Residual solvents:complying
- Refractive Index:n20/D 1.329 (lit.)
- Explosion Limit:36 %
Frequently Bought Together