5-Bromopentyl acetate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15848-22-3
:
Formula:C₇H₁₃BrO₂
MDL Number:MFCD00039204
:209.08 g/mol
Boiling Pt:109…110 °C (15 mmHg)
Density:1.255
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:239-961-8
Related Information
Frequently Bought Together