
2-(2-Ethoxyethoxy)ethyl acetate, Sigma-Aldrich®
Catalog # 8.14635.1000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-25 °C
- Molar Mass:176.21 g/mol
- Boiling Point:217 - 218 °C
- Vapor Pressure:0.3 hPa (20 °C)
- Flash Point:100 °C
- Explosion Limit:1.5 %(V)
- Density:1.01 g/cm3 (20 °C)
- pH:(H2O, 20 °C) neutral
- Ignition Point:310 °C
Specification Test Results
About this item
:
:176.21 g/mol
Boiling Pt:210 °C (1013 hPa)
Melting Pt:–25 °C
Density:1.0114 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:203-940-1
:GHS07
CAS Number:112-15-2
Formula:C₈H₁₆O₄
MDL Number:MFCD00041928